missing translation for 'onlineSavingsMsg'
Learn More

Yttrium Standard For ICP, 2% HNO3 in 1000μg/mL, Assurance Grade, SPEX CertiPrep™

Product Code. 10661563 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
Unit Size:
125mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10661563 125 mL Triple-leached LDPE bottle 125mL
10102682 500 mL Triple-leached LDPE bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10661563 Supplier SPEX Certiprep Supplier No. PLY22Y
  • ONLINE EXCLUSIVE PRICE
    74.70 €
    58.93 € / 125mL
    Save 15.77 €
    21% Off
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Y1000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 7440-65-5 , 7697-37-2 , 7732-18-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]

Specifications

Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
CAS Min % 1.0, 5.0, 94.0
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 1.0, 5.0, 94.0
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
Solvent or Matrix 2% HNO3
Quantity 125 mL
Grade Plasma
Concentration or Composition (by Analyte or Components) Matrix    Concentration    Cat. No.
2-5% HNO3    1000μg/mL    PLY2-2Y, -2T, -2X    10,000    PLY2-3Y, -3X
Chemical Name or Material Yttrium Standard
Show More Show Less
compliance-icons
Product Identifier
  • 1000
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.