Learn More
YY1 Mouse anti-Human, Mouse, Non-human primate, Rat, Clone: 2E11C5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66281-1-IG-150UL
Product Code. 16818015
  • 189.00 € / 150µL
Description
YY1 is a ubiquitously distributed transcription factor belonging to the GLI-Kruppel class of zinc finger proteins. The protein is involved in repressing and activating a diverse number of promoters. YY1 may direct histone deacetylases and histone acetyltransferases to a promoter in order to activate or repress the promoter, thus implicating histone modification in the function of YY1.
Specifications
Specifications
YY1
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P25490, Q00899
YY1
YY1 Fusion Protein Ag17732
150 μL
Primary
Human, Mouse, Non-human Primate, Rat
Antibody
IgG2a
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
2E11C5
Unconjugated
YY1
DELTA, Delta transcription factor, INO80 complex subunit S, INO80S, NF E1, UCRBP, Yin and yang 1, YIN YANG 1, YY 1, YY1, YY1 transcription factor
Mouse
Protein A
RUO
22632, 24919, 7528
-20°C