missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Metominostrobin, TRC

Product Code. 30135504
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135504 25 mg 25mg
30132886 100 mg 100mg
30006161 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30135504 Supplier Toronto Research Chemicals Supplier No. TRCM33875825MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133408-51-2
Molecular Formula C16 H16 N2 O3
Molecular Weight (g/mol) 284.31
Synonym Benzeneacetamide, α-(methoxyimino)-N-methyl-2-phenoxy-, (αZ)- (9CI, ACI), (αZ)-α-(Methoxyimino)-N-methyl-2-phenoxybenzeneacetamide (ACI), Benzeneacetamide, α-(methoxyimino)-N-methyl-2-phenoxy-, (Z)- (ZCI), (Z)-2-(Methoxyimino)-N-methyl-2-(2-phenoxyphenyl)acetamide, (Z)-Metominostrobin, (Z)-N-Methyl-2-(2-phenoxyphenyl)-2-(methoxyimino)acetamide
IUPAC Name (2Z)-2-methoxyimino-N-methyl-2-(2-phenoxyphenyl)acetamide
SMILES CNC(=O)\C(=N/OC)\c1ccccc1Oc2ccccc2

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H16N2O3/c1-17-16(19)15(18-20-2)13-10-6-7-11-14(13)21-12-8-4-3-5-9-12/h3-11H,1-2H3,(H,17,19)/b18-15-
Formula Weight 284.1161
Chemical Name or Material (Z)-Metominostrobin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.