missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Thiothixene, MedChemExpress

Product Code. 16494178
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494178 10 mg 10mg
16466897 5 mg 5mg
16412986 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16494178 Supplier Medchem Express Supplier No. HY108324
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress (Z)-Thiothixene is an antagonist of serotonergic receptor extracted from patent US 20150141345 A1.

(Z)-Thiothixene is an antagonist of serotonergic receptor extracted from patent US 20150141345 A1.

Specifications

Chemical Name or Material (Z)-Thiothixene
CAS 3313-26-6
Color White
Purity Grade Notes Research
Molecular Formula C23H29N3O2S2
Quantity 10 mg
Solubility Information DMSO : 10 mg/mL (22.54 mM; ultrasonic and warming and heat to 60°C)
SMILES O=S(C1=CC(/C2=C\CCN3CCN(C)CC3)=C(SC4=C2C=CC=C4)C=C1)(N(C)C)=O
Molecular Weight (g/mol) 443.63
Formula Weight 443.63
Percent Purity 99.09%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.