missing translation for 'onlineSavingsMsg'
Learn More

Z433927330, MedChemExpress

Product Code. 16477977
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16477977 5 mg 5mg
16455926 100 mg 100mg
16421477 50 mg 50mg
16415268 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16477977 Supplier Medchem Express Supplier No. HY126074
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Z433927330 is a potent and selective inhibitor of Aquaporin-7 (AQP7), less potently inhibits AQP3 and AQPs9, with IC50s of ∼0.2 μM, ∼0.7 μM and ∼1.1 μM for mAQP7, mAQP3 and mAQP9, respectively.

Z433927330 is a potent and selective inhibitor of Aquaporin-7 (AQP7), less potently inhibits AQP3 and AQPs9, with IC50s of ∼0.2 μM, ∼0.7 μM and ∼1.1 μM for mAQP7, mAQP3 and mAQP9, respectively.

Specifications

Chemical Name or Material Z433927330
Physical Form Solid
CAS 1005883-72-6
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H20N4O3
Solubility Information DMSO : ≥ 280 mg/mL (768.39 mM)
SMILES O=C(OCC)C1=CC=C(NC(NCC2=CC=C(N3N=CC=C3)C=C2)=O)C=C1
Molecular Weight (g/mol) 364.4
Formula Weight 364.4
Percent Purity 98.01%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.