Learn More
ZA20D3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-90060
This item is not returnable.
View return policy
Description
ZA20D3 Polyclonal specifically detects ZA20D3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| ZA20D3 | |
| Immunohistochemistry (Paraffin) | |
| Polyclonal | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |