missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZAG Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97955-50ul
This item is not returnable.
View return policy
Description
ZAG Polyclonal antibody specifically detects ZAG in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ZAG | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:5000 | |
| alpha-2-glycoprotein 1, zinc, alpha-2-glycoprotein 1, zinc-binding, ZA2G, ZAGzn-alpha-2-GP, zinc, zinc-alpha-2-glycoprotein, zn-alpha-2-glycoprotein, ZNGP1 | |
| Produced in rabbits immunized with E. coli-derived Human ZAG fragment. | |
| 50 μg | |
| Endocrinology, Immunology, Innate Immunity, Signal Transduction | |
| 563 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction