missing translation for 'onlineSavingsMsg'
Learn More

Zardaverine, MedChemExpress

Product Code. 16472657
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16472657 50 mg 50mg
16456568 10 mg 10mg
16409147 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16472657 Supplier Medchem Express Supplier No. HY15485
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Zardaverine is a newly developed dual-selective PDE3/4 inhibitor with IC50 values of 0.5 uM and 0.8 uM respectively.

Zardaverine is a newly developed dual-selective PDE3/4 inhibitor with IC50 values of 0.5 uM and 0.8 uM respectively.

Specifications

Chemical Name or Material Zardaverine
CAS 101975-10-4
Color White
Purity Grade Notes Research
Molecular Formula C12H10F2N2O3
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (93.21 mM; Need ultrasonic)
SMILES O=C1C=CC(C2=CC=C(OC(F)F)C(OC)=C2)=NN1
Molecular Weight (g/mol) 268.22
Formula Weight 268.22
Percent Purity 98.16%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.