missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZC3H15 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
214.00 € - 493.00 €
Specifications
| Antigen | ZC3H15 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18665041
|
Novus Biologicals
NBP2-94541-0.02ml |
0.02 mL |
214.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18668261
|
Novus Biologicals
NBP2-94541-0.1ml |
0.1 mL |
493.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZC3H15 Polyclonal antibody specifically detects ZC3H15 in Human, Mouse samples. It is validated for Western BlotSpecifications
| ZC3H15 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| DFRP1, DRG family-regulatory protein 1, HT010, LEREPO4MSTP012, Likely ortholog of mouse immediate early response erythropoietin 4, zinc finger CCCH domain-containing protein 15, zinc finger CCCH-type containing 15 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ZC3H15 (NP_060941.2). MPPKKQAQAGGSKKAEQKKKEKIIEDKTFGLKNKKGAKQQKFIKAVTHQVKFGQQNPRQVAQSEAEKKLKKDDKKKELQELNELFKPVVAAQKISKGADPKSVVCAFFKQGQCTKGDKCKFSHDLTLERKCEKRSVYIDARDEELEKDTMDNWDEKKLEEVVNKKHGEAEKKKPKTQIVCKHFLEAIENNKYGWFWVCPGGGDICMYRHALPPGFVLKKDKKKEEKEDEISLEDLIERERSALGPNVTKITLESFLAWKKRKRQEKIDKLEQDMERRKADFKAGKALVISGREVFEFRPE | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 55854 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title