missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZC3HAV1 Polyclonal Antibody

Product Code. 12626213
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12626213 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12626213 Supplier Invitrogen™ Supplier No. PA520986

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Hela cell lysate. PA5-20986 can be used with blocking peptide PEP-1100.

The zinc finger antiviral protein (ZC3HAV1) is a CCCH type zinc finger protein that induces an innate immunity to infections by retrovirus by preventing the accumulation of viral RNAs in the cytoplasm and recruits the RNA processing exosome to degrade target RNAs, thereby inhibiting virus replication. ZC3HAV1 is localized in the cytoplasm at steady state, but shuttles between nucleus and cytoplasm in a XPO1-dependent manner. ZAP is a direct target gene of IRF3 action in cellular antiviral responses.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZC3HAV1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Zc3hav1
Gene Accession No. Q7Z2W4
Gene Alias 1200014N16Rik; 2900058M19Rik; 9130009D18Rik; 9830115L13Rik; ADP-ribosyltransferase diphtheria toxin-like 13; ARTD13; CCCH-type zinc finger antiviral protein (Zap); D6Bwg1452e; FLB6421; Inactive Poly [ADP-ribose] polymerase 13; PARP13; PRO1677; rZAP; ZAP; ZC3H2; Zc3hav1; ZC3HDC2; zinc finger antiviral protein; Zinc finger CCCH domain-containing protein 2; zinc finger CCCH type, antiviral 1; zinc finger CCCH-type antiviral protein 1; zinc finger CCCH-type containing, antiviral 1; zinc finger CCCH-type, antiviral 1
Gene Symbols Zc3hav1
Host Species Rabbit
Immunogen A 14 amino acid peptide near the carboxy terminus of human ZC3HAV1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56829
Target Species Human
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.