missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCCHC11/TUT4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-83027
This item is not returnable.
View return policy
Description
ZCCHC11/TUT4 Polyclonal specifically detects ZCCHC11/TUT4 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ZCCHC11/TUT4 | |
| Polyclonal | |
| Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
| DKFZp779C1943, EC 2.7.7.52, KIAA0191FLJ42878, PAP associated domain containing 3, PAPD3, terminal uridylyltransferase 4, TUT4, TUTase 4, Zinc finger CCHC domain-containing protein 11, zinc finger, CCHC domain containing 11 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TUT4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:CNCIDLSKSPDPDKSTGTDCRSNLETESSHQSVCTDTSATSCNCKATEDASDLNDDDNLPTQELYYVFDKFILTSGK | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 23318 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction