missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZCWPW2 Antibody (OTI10A12), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
256.00 € - 550.00 €
Specifications
| Antigen | ZCWPW2 |
|---|---|
| Clone | OTI10A12 |
| Dilution | Western Blot 1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30306485
|
Novus Biologicals
NBP3-44988-100ul |
100 μL |
550.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30306689
|
Novus Biologicals
NBP3-44988-30ul |
30 μL |
256.00 €
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZCWPW2 Monoclonal antibody specifically detects ZCWPW2 in Human samples. It is validated for Western BlotSpecifications
| ZCWPW2 | |
| Western Blot 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| 152098 | |
| IgG1 | |
| >95% | |
| Antibody |
| OTI10A12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| ZCW2, zinc finger CW-type PWWP domain protein 2, zinc finger, CW type with PWWP domain 2 | |
| Full length human recombinant protein of human ZCWPW2 (NP_001035522) produced in E.coli. | |
| Primary | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title