missing translation for 'onlineSavingsMsg'
Learn More

ZD7288, MedChemExpress

Product Code. 16463338
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
missing translation for 'unitSize'
10mg
5mg
This item is not returnable. View return policy

Product Code. 16463338

missing translation for 'mfr': Medchem Express HY101346

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ZD7288 (ICI D7288) is a selective hyperpolarization-activated cyclic nucleotide-gated (HCN) channel blocker.

ZD7288 (ICI D7288) is a selective hyperpolarization-activated cyclic nucleotide-gated (HCN) channel blocker.

Specifications

Chemical Name or Material ZD7288
Physical Form Solid
CAS 133059-99-1
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H21ClN4
For Use With (Application) Neuroscience-Neuromodulation
Synonym ICI D7288
Solubility Information DMSO : 50 mg/mL (170.76 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (170.76 mM)
SMILES [H]Cl.C/N=C1C=C(N(CC)C2=CC=CC=C2)N=C(C)N/1C
Molecular Weight (g/mol) 292.81
Formula Weight 292.81
Percent Purity 99.86%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.