missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHC3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553460
This item is not returnable.
View return policy
Description
Immunogen sequence: FLHCFEEDWT TYGLNREEMA ETGISLHEKM QPLNFSSTEC SSFSPPTT Highest antigen sequence identity to the following orthologs: Mouse - 40%, Rat - 40%.
Golgi-localized palmitoyltransferase that catalyzes the addition of palmitate onto various protein substrates (PubMed:19001095, PubMed:21926431, PubMed:22240897, PubMed:23034182, PubMed:22314500). Plays an important role in G protein-coupled receptor signaling pathways involving GNAQ and potentially other heterotrimeric G proteins by regulating their dynamic association with the plasma membrane (PubMed:19001095). Palmitoylates ITGA6 and ITGB4, thereby regulating the alpha-6/beta-4 integrin localization, expression and function in cell adhesion to laminin (PubMed:22314500). Plays a role in the TRAIL-activated apoptotic signaling pathway most probably through the palmitoylation and localization to the plasma membrane of TNFRSF10A (PubMed:22240897). In the brain, by palmitoylating the gamma subunit GABRG2 of GABA(A) receptors and regulating their postsynaptic accumulation, plays a role in synaptic GABAergic inhibitory function and GABAergic innervation. Palmitoylates the neuronal protein GAP43 which is also involved in the formation of GABAergic synapses. Palmitoylates NCDN thereby regulating its association with endosome membranes. Probably palmitoylates PRCD and is involved in its proper localization within the photoreceptor. Could mediate the palmitoylation of NCAM1 and regulate neurite outgrowth. Could palmitoylate DNAJC5 and regulate its localization to Golgi membranes. Also constitutively palmitoylates DLG4. May also palmitoylate SNAP25. Could palmitoylate the glutamate receptors GRIA1 and GRIA2 but this has not been confirmed in vivo. Could also palmitoylate the D(2) dopamine receptor DRD2 (PubMed:26535572). [UniProt].
Specifications
| ZDHC3 | |
| Polyclonal | |
| Unconjugated | |
| ZDHHC3 | |
| 1110020O22Rik; 1810006O10Rik; 2210017C02Rik; DHHC1 protein; DHHC-3; GABA-A receptor-associated membrane protein 1; Godz; Golgi apparatus-specific protein with DHHC zinc finger domain; golgi-specific DHHC Zinc Finger Protein; Gramp1; HSD49; Palmitoyltransferase ZDHHC3; Protein DHHC1; Zdhhc3; Zfp373; Zinc finger DHHC domain-containing protein 3; zinc finger DHHC-type containing 3; zinc finger protein 373; zinc finger, DHHC domain containing 3; zinc finger, DHHC-type containing 3; ZNF373 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51304 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NYG2 | |
| ZDHHC3 | |
| Recombinant protein corresponding to Human ZDHC3. Recombinant protein control fragment (Product #RP-91170). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction