Learn More
ZEB2 Antibody (CL13467) [Alexa Fluor™ 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-24559AF488
Product Code. 30293048
  • 737.00 € / 0.10mL
Description
ZEB2 Monoclonal antibody specifically detects ZEB2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZEB2
Monoclonal
Alexa Fluor 488
KIAA0569FLJ42816, SIP1HSPC082, Smad-interacting protein 1, SMADIP1ZFHX1BSIP-1, ZFX1B, zinc finger E-box binding homeobox 2, zinc finger E-box-binding homeobox 2, zinc finger homeobox 1b, Zinc finger homeobox protein 1b
This antibody was developed using a recombinant protein derived from O60315, with the exact immunogen sequence remaining proprietary.
0.1 mL
Core ESC Like Genes, Neuroscience, Stem Cell Markers
9839
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL13467
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified