missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zebrafish Basolateral Pole of Cells Antibody (FIS 2H9/1) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
190.00 € - 351.00 €
Specifications
| Antigen | Zebrafish Basolateral Pole of Cells |
|---|---|
| Clone | FIS 2H9/1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18615907
|
Novus Biologicals
NBP2-50370-0.025ml |
0.025 mL |
190.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18633409
|
Novus Biologicals
NBP2-50370 |
0.1 mL |
351.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Zebrafish Basolateral Pole of Cells Monoclonal antibody specifically detects Zebrafish Basolateral Pole of Cells in Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Zebrafish Basolateral Pole of Cells | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Developmental Biology, Epitope Tags | |
| PBS | |
| IgG1 | |
| Protein A purified |
| FIS 2H9/1 | |
| Western Blot 1:100 - 1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Zebrafish | |
| Lysate of zebrafish intestine | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title