missing translation for 'onlineSavingsMsg'
Learn More

Zephirol, TRC

Product Code. 17974945
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17974945 50 mg 50mg
30138675 10 g 10g
30138402 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17974945 Supplier Toronto Research Chemicals Supplier No. TRCZ34300050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 139-07-1
Formula Weight 339.2693
InChI Formula InChI=1S/C21H38N.ClH/c1-4-5-6-7-8-9-10-11-12-16-19-22(2,3)20-21-17-14-13-15-18-21;/h13-15,17-18H,4-12,16,19-20H2,1-3H3;1H/q+1;/p-1
IUPAC Name benzyl-dodecyl-dimethylazanium;chloride
Molecular Formula C21 H38 N . Cl
Molecular Weight (g/mol) 339.99
Percent Purity >95%
SMILES [Cl-].CCCCCCCCCCCC[N+](C)(C)Cc1ccccc1
Synonym Benzenemethanaminium, N-dodecyl-N,N-dimethyl-, chloride (1:1), Ammonium, benzyldodecyldimethyl-, chloride (8CI), Benzenemethanaminium, N-dodecyl-N,N-dimethyl-, chloride (9CI), Benzyldodecyldimethylammonium chloride (6CI), Accelerator 1227, BAC 12, BTC 1218-50, Bas 2631, Benzododecinium chloride, BenzylQUAT, Benzyldimethyldodecylammonium chloride, Benzyldimethyllaurylammonium chloride, Benzyllauryldimethylammonium chloride, Catigene OM, Catinal CB 50, Catiogen BC 50, Catiogen PAN, Catiolite BC 50, Cationic 1227, Cationic Surfactant 1227, Cequartyl A, Croda N-Dodecyl, D 1227, DYK 1125, Dehyquart LDB, Dimethylaurylbenzylammonium chloride, Dimethylbenzyldodecylammonium chloride, Dimethylbenzyllaurylammonium chloride, Dimethyldodecylbenzylammonium chloride, Dimethyllaurylbenzylammonium chloride, Dodecylbenzyldimethylammonium chloride, Dodecyldimethylbenzylammonium chloride, Germicide 1227, Katamin AB 18, Lauralkonium chloride, Lauryl benzalkonium chloride, Laurylbenzyldimethylammonium chloride, Lauryldimethylbenzylammonium chloride, Leveling Agent 1227, Loraquat B 50, N-Benzyl-N,N-dimethyl-N-laurylammonium chloride, N-Benzyl-N-dodecyl-N,N-dimethylammonium chloride, N-Dodecyl-N,N-dimethyl-N-benzylammonium chloride, N-Dodecyl-N,N-dimethylbenzenemethanaminium chloride, N-Lauryl-N,N-dimethyl-N-benzylammonium chloride, Noramium DA 50, Orthosan HM, Pionin B 231, QBA 1211, QBA 1211S, Retarder N, Rewoquat B 50, Rolcril, Surfactant 1227, Swanol CA 100, Swanol CA 101, Tetranil BC 80, Texnol R 5, Vantoc CL, Zephirol
Chemical Name or Material Benzyldimethyldodecylammonium Chloride
Grade HPLC
Show More Show Less

Specifications

Quantity 50 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.