Learn More
Isofield™ Zetta 230 Non-Sterile TieOon Cleanroom Facemask - ISO Class 4/5 Compatible
BIOPHARMA
Product Code. 30178905
Change view
Click to view available options
Size:
Wide
Unit Size:
Pack of 500
Product Code. Size Unit Size
30178905 Wide Pack of 500
1 options
Product Code. 30178905 Supplier Isofield™ Supplier No. 60330
Estimated Shipment: 18-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
The Zetta is ISO Class 4 compatible. The Zetta is also made from low-linting cleanroom grade material to ensure best in class cleanliness. The individually packed Zetta has excellent bacterial and particle filtration efficiency, and an excellent choice for GMP Grade C/D environments.
  • Non-sterile tie-on cleanroom facemask
  • 230 mm length with malleable nose strip
  • 3-layer non-woven configuration
  • Highly efficient bacterial and particle filtration
  • Particle Shedding (Helmke Drum) Category I
  • Individually packed to maintain low CFU count
  • Pouches printed with IPA resistant ink to maintain traceability
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Ties
Material Polyethylene, Polypropylene
Size Wide
Sterility Non-sterile
Style Pleated