missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZFP28 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-81408
This item is not returnable.
View return policy
Description
ZFP28 Polyclonal specifically detects ZFP28 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ZFP28 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| DKFZp686K03205, KIAA1431, krueppel-like zinc finger factor X6, MGC104540, mkr5, zfp-28, zinc finger factor X6, zinc finger protein 28 homolog, zinc finger protein 28 homolog (mouse) | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 140612 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| ZFP28 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LLEQEKEPWMVKRELTGSLFSGQRSVHETQELFPKQDSYAEGVTDRTSNTKLDCSSFRENWDSDYVFGRKLAVGQETQFRQEPITHNKTLSKERERTYNKSGRWFYLDDSEEKVHNRDSIKNFQKSSVVIKQTGIYAG | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction