Learn More
ZFP36L1 Mouse, anti-Human, CoraLite Plus 488, Clone: 1H6G6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68038-20UL
Description
The ZFP36L1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human ZFP36L1. This antibody recognizes human, rat antigen. The ZFP36L1 antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™ plus 488.
Specifications
| ZFP36L1 | |
| 1H6G6 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Rat | |
| 155 kDa | |
| IgG2b |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| 1000 μg/mL | |
| Berg36, BRF1, Butyrate response factor 1, cMG1, EGF response factor 1, ERF 1, ERF1, Protein TIS11B, RNF162B, TIS11B, ZFP36L1 | |
| 20 μL | |
| 677 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |