missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZFX Polyclonal Antibody
GREENER_CHOICE

Product Code. 16366934
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16366934 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16366934 Supplier Invitrogen™ Supplier No. PA578234

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: K562, U87-MG, SK-N-SH, IMR32, SK-N-AS Predicted Reactivity: Mouse (84%), Rat (84%), Bovine (86%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZFX is a member of the Kruppel C2H2-type zinc-finger protein family. The full-length protein contains an acidic transcriptional activation domain (AD), a nuclear localization sequence (NLS) and a DNA binding domain (DBD) consisting of 13 C2H2-type zinc fingers. Studies in mouse embryonic and adult hematopoietic stem cells showed that this gene was required as a transcriptional regulator for self-renewal of both stem cell types, but it was dispensable for growth and differentiation of their progeny.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFX
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 2.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene ZFX
Gene Accession No. P17010
Gene Alias RGD1562664; X-linked zinc finger protein; Zfx; Zfx5; Zfx5,6; Zfx6; zinc finger protein X-linked; zinc finger protein X-linked 5; zinc finger protein X-linked 6; zinc finger protein, X-linked; zinc finger X-chromosomal protein; ZNF926
Gene Symbols ZFX
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus of human ZFX. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7543
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.