missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZFYVE9 Recombinant Rabbit Monoclonal Antibody (K01_2P39)

Product Code. 30829334
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829334 100 μL
30828103 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829334 Supplier Invitrogen™ Supplier No. MA564201
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

ZFYVE9 Recombinant Monoclonal Antibody for IHC (P), Western Blot

Early endosomal protein that functions to recruit SMAD2/SMAD3 to intracellular membranes and to the TGF-beta receptor. Plays a significant role in TGF-mediated signaling by regulating the subcellular location of SMAD2 and SMAD3 and modulating the transcriptional activity of the SMAD3/SMAD4 complex. Possibly associated with TGF-beta receptor internalization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZFYVE9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2P39
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene Zfyve9
Gene Accession No. O95405
Gene Alias E030027L17; hSARA; MAD, mothers against decapentaplegic homolog interacting protein, receptor activation anchor; MADH-interacting protein; MADHIP; Mothers against decapentaplegic homolog-interacting protein; Novel serine protease; NSP; PPP1R173; protein phosphatase 1, regulatory subunit 173; Receptor activation anchor; SARA; Smad anchor for receptor activation; SMADIP; ZFYVE9; Zinc finger FYVE domain-containing protein 9; zinc finger FYVE-type containing 9; zinc finger, FYVE domain containing 9
Gene Symbols Zfyve9
Host Species Rabbit
Immunogen A synthetic peptide of human SARA.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9372
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.