missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Zika Virus Envelope (strain Zika SPH2015) Recombinant Rabbit Monoclonal Antibody (033), HRP
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

189.00 € - 569.00 €

Specifications

Antigen Zika Virus Envelope (strain Zika SPH2015)
Clone 033
Concentration 1 mg/mL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Applications ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30536104
GREENER_CHOICE
View Documents
Invitrogen™
MA560379
100 μg
569.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30535119
View Documents
Invitrogen™
MA560380
20 μg
189.00 €
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Zika Virus Envelope (strain Zika SPH2015) Recombinant Monoclonal Antibody for ELISA

As a member of the Flaviviridae family, ZIKV has a single positive sense RNA genome, which is initially translated as a single polyprotein which is then cleaved post-transitionally into three structural proteins (C, PrM or M, and E) and seven nonstructural (NS) proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B and NS5)(1). The glycosated NS1 forms a homodimer inside the cells and is necessary for viral replication and late in infection (2). NS1 is also secreted into the extracellular space as a hexameric lipoprotein particle, which assists the virus in immune evasion and pathogenesis by interacting with components from both innate and adaptive immune systems as well as other host factors (1). Reference 1) Song, H.; Qi, J.; Haywood, J.; Shi, Y.; Gao, G.F. (2016). Nat. Struct. Mol. Biol. 23, 456-458. 2) Suthar, M.S., Diamond, M.S. and Gale, M. Jr. (2013). Nat. Rev. Microbiol. 11, 115-128.
Specifications

Specifications

Zika Virus Envelope (strain Zika SPH2015)
1 mg/mL
ELISA
HRP
Rabbit
RUO
PBS with no preservative
Recombinant Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-E/Envelope protein (Domain III) protein (Val593-Lys699).
Antibody
033
-20°C, Avoid Freeze/Thaw Cycles
Recombinant Monoclonal
Liquid
IgG
Virus
Zika Envelope
Primary
Protein A
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.