Learn More
Invitrogen™ Zika Virus NS1 (strain Zika SPH2015) Recombinant Rabbit Monoclonal Antibody (201)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA560381
 View more versions of this product
Product Code. 30534108
  • 429.00 € / 100µL
Description
Zika Virus NS1 (strain Zika SPH2015) Recombinant Monoclonal Antibody for ELISA
As a member of the Flaviviridae family, ZIKV has a single positive sense RNA genome, which is initially translated as a single polyprotein which is then cleaved post-transitionally into three structural proteins (C, PrM or M, and E) and seven nonstructural (NS) proteins (NS1, NS2A, NS2B, NS3, NS4A, NS4B and NS5)(1). The glycosated NS1 forms a homodimer inside the cells and is necessary for viral replication and late in infection (2). NS1 is also secreted into the extracellular space as a hexameric lipoprotein particle, which assists the virus in immune evasion and pathogenesis by interacting with components from both innate and adaptive immune systems as well as other host factors (1). Reference 1) Song, H.; Qi, J.; Haywood, J.; Shi, Y.; Gao, G.F. (2016). Nat. Struct. Mol. Biol. 23, 456-458. 2) Suthar, M.S., Diamond, M.S. & Gale, M. Jr. (2013). Nat. Rev. Microbiol. 11, 115-128.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Zika Virus NS1 (strain Zika SPH2015)
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Rabbit
Protein A
RUO
Virus
Antibody
IgG
ELISA
201
Unconjugated
Zika NS1; ZIKV NS1; ZIKV-NS1
Recombinant Zika virus (ZIKV) (strain Zika SPH2015) ZIKV-NS1 protein (Val796-Leu1157).
100 μL
Primary
-20°C, Avoid Freeze/Thaw Cycles