missing translation for 'onlineSavingsMsg'
Learn More

Zinc chloride, 1.0M solution in diethyl ether, AcroSeal™

Product Code. 10292572 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
800 mL
Unit Size:
100mL
800mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10292572 100 mL 100mL
10029051 800 mL 800mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10292572 Supplier Thermo Scientific Acros Supplier No. 370051000
  • ONLINE EXCLUSIVE PRICE
    90.95 €
    88.30 € / 100mL
    Save 2.65 €
    2% Off
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7646-85-7 | Cl2Zn | 136.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Zinc chloride
Color Blue-Gray or Colorless to Yellow
Name Note 1.0M solution in diethyl ether
CAS 7646-85-7
Molecular Formula Cl2Zn
Linear Formula ZnCl2
MDL Number MFCD00011295
Concentration 0.9 to 1.1M
Fieser 01,1289; 02,464; 03,338; 05,763; 06,676; 08,536; 09,522; 10,461; 11,602; 12,446; 13,44; 14,177; 15,89; 16,391
Density 0.8400g/mL
Merck Index 15, 10331
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
SMILES Cl[Zn]Cl
IUPAC Name dichlorozinc
Molecular Weight (g/mol) 136.28
PubChem CID 5727
ChEBI CHEBI:49976
Formula Weight 136.29
Packaging AcroSeal™ Glass Bottle
Quantity 100 mL
Physical Form Solution, May Contain A Small Amount Precipitate
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc chloride, 1.0M solution in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.