missing translation for 'onlineSavingsMsg'
Learn More

Zinc meso-tetraphenylporphine

Product Code. 11358358
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358358 2 g 2g
11368358 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358358 Supplier Thermo Scientific Alfa Aesar Supplier No. 039552.04
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14074-80-7 | C44H28N4Zn | 678.12 g/mol

A porphyrin used in light emitting diodes. It has valuable biological applications in molecular biology, fluoroimmunoassay and new pharmaceutical developments.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14074-80-7
Molecular Formula C44H28N4Zn
Molecular Weight (g/mol) 678.12
MDL Number MFCD00012155,MFCD00012155
InChI Key FGEZGTKFMCNEOZ-UHFFFAOYSA-N
Synonym zinc meso-tetraphenylporphine
PubChem CID 91667917
IUPAC Name 5,10,15,20-tetraphenylporphyrin-22,24-diide;zinc
SMILES [Zn].[N-]1C2=CC=C1C(=C1C=CC(=N1)C(=C1[N-]C(C=C1)=C(C1=NC(C=C1)=C2C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Purple
Odor Odorless
Quantity 2 g
Solubility Information Insoluble in water.
Formula Weight 678.1
Physical Form Crystalline
Chemical Name or Material Zinc meso-tetraphenylporphine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.