Learn More
ZMAT4 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24137-1-AP-150UL
This item is not returnable.
View return policy
Description
The ZMAT4 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ZMAT4. This antibody recognizes human, mouse antigen. The ZMAT4 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| ZMAT4 | |
| 800 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| zinc finger, matrin type 4, ZMAT4 | |
| 150 μL | |
| 79698 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |