missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ZNF23 May be involved in transcriptional regulation. May have a role in embryonic development. Belongs to the krueppel C2H2-type zinc-finger protein family.
Specifications
Specifications
| Antigen | ZNF23 |
| Applications | Western Blot, Immunocytochemistry, Immunoprecipitation, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 0.32 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ZNF23 |
| Gene Accession No. | P17027 |
| Gene Alias | KOX16, Zfp612, Zinc finger protein 23, Zinc finger protein 359, Zinc finger protein KOX16, ZNF23, ZNF359, ZNF612 |
| Gene Symbols | Zfp612, ZNF23 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?