missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF274 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15524122
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15524122 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15524122 Supplier Invitrogen™ Supplier No. PA540385

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: YPELQLDPKL DPLPAESPLM NIEVVEVLTL NQEVAGPRNA QIQALYAEDG Sequence homology: Cow: 93%; Dog: 93%; Human: 100%; Mouse: 86%; Pig: 86%; Rabbit: 100%; Rat: 86%.

ZNF274 is a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus.This gene encodes a zinc finger protein containing five C2H2-type zinc finger domains, one or two Kruppel-associated box A (KRAB A) domains, and a leucine-rich domain. The encoded protein has been suggested to be a transcriptional repressor. It localizes predominantly to the nucleolus. Alternatively spliced transcript variants encoding different isoforms exist. These variants utilize alternative polyadenylation signals.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF274
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF274
Gene Accession No. Q96GC6
Gene Alias DKFZp686K08243; HFB101; KRAB zinc finger protein HFB101; neurotrophin receptor-interacting factor homolog; SP2114; Zf2; zinc finger protein 274; Zinc finger protein HFB101; zinc finger protein with KRAB and SCAN domains 19; Zinc finger protein zfp2; ZKSCAN19; ZNF274; ZSCAN51
Gene Symbols ZNF274
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF274 (aa 87-136).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10782
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.