missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: FPVAEQDKLL KHMIIEHKIV IADVKLVADF QRYILYWRKR FTEQPITDFC SVIRINSTAP FEEQENYFLL CDVLPEDRIL REELQKQRLR EILEQQQQER NDTNFHGVCM FCNEEFLGNR SVILNH Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 93%.
ZNF277 (Zinc Finger Protein 277) is a Protein Coding gene. Diseases associated with ZNF277 includeSpecific Language ImpairmentandSpeech And Communication Disorders. Gene Ontology (GO) annotations related to this gene includeRNA polymerase II core promoter sequence specific DNA binding.
Specifications
Specifications
| Antigen | ZNF277 |
| Applications | ChIP Assay, Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.20 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | ZNF277 |
| Gene Accession No. | Q9NRM2 |
| Gene Alias | NRIF4; Nuclear receptor-interacting factor 4; zinc finger protein (C2H2 type) 277; zinc finger protein 277; zinc finger protein 277 pseudogene; ZNF277; ZNF277P |
| Gene Symbols | ZNF277 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
