missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF297B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
280.00 € - 572.00 €
Specifications
| Antigen | ZNF297B |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18421041
|
Novus Biologicals
NBP1-92621-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18089276
|
Novus Biologicals
NBP1-92621 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF297B Polyclonal specifically detects ZNF297B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| ZNF297B | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 23099 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:GVEEDFHIGEKKVEAEFDEQADESNYDEQVDFYGSSMEEFSGERSDGNLIGHRQEAALAAGYSENIEMVTGIKEEASHLGFSATDKLYPCQCGKSFTHKSQRDRHMSMHLGL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| KIAA0414, ZBTB22BZNF-X, zinc finger and BTB domain containing 43, Zinc finger and BTB domain-containing protein 22B, zinc finger and BTB domain-containing protein 43, Zinc finger protein 297BFLJ22470, ZNF297B, ZnF-x | |
| ZBTB43 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title