missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF318 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
292.00 € - 572.00 €
Specifications
| Antigen | ZNF318 |
|---|---|
| Dilution | Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18436120
|
Novus Biologicals
NBP1-83209 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18447010
|
Novus Biologicals
NBP1-83209-25ul |
25 μL |
292.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ZNF318 Polyclonal antibody specifically detects ZNF318 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| ZNF318 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research | |
| PBS (pH 7.2) and 40% Glycerol | |
| 24149 | |
| IgG | |
| Immunogen affinity purified |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Zinc finger and SCAN domain-containing protein 15, zinc finger protein 397, Zinc finger protein 47ZSCAN15MGC13250, ZNF47 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: IENKGTMVETALKEPQGNLYQWGPLPGIPKDNSPLREKFGSFLCHKDNLDLKAEGPERHTDFLLPHERASQDGSGFSRILSMLADSTSTQEKRRRSFPDIEDEEKFLYGDEEEDLKAESVPKP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title