missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF346 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-38437-100ul
This item is not returnable.
View return policy
Description
ZNF346 Polyclonal antibody specifically detects ZNF346 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifica
| ZNF346 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| KIAA0844ZNF365D, MGC41821, MGC87345, Su48, talanin, UANprotein ZNF365, zinc finger protein 365 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-294 of human ZNF346 (NP_036411.1).,, Sequence:, MEYPAPATVQAADGGAAGPYSSSELLEGQEPDGVRFDRERARRLWEAVSGAQPVGREEVEHMIQKNQCLFTNTQCKVCCALLISESQKLAHYQSKKHANKVKRYLAIHGMETLKGETKKLDSDQKSSRSKDKNQCCPICNMTFSSPVVAQSHYLGKTHAKNLKLKQQSTKVEALHQNREMIDPDKFCSLCHATFNDPVMAQQHYVGKKHRKQETKLKLMARYGRLADPAVTDFPAGKGYPCKTCKIVLNSIEQYQAHVSGFKHKNQSPKTVASSLGQIPMQRQPIQKDSTTLED | |
| 100 μL | |
| Apoptosis | |
| 23567 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto