missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF358 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15524662
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15524662 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15524662 Supplier Invitrogen™ Supplier No. PA541155

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: LILDPNSDTL SPGDPKVDPI SSGLTATPQV LATSPAVLPA PASPPRPFSC Sequence homology: Human: 100%.

ZNF358, a gene located on chromosome 19, encodes a protein involved in embryogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF358
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF358
Gene Accession No. Q9NW07
Gene Alias FLJ10390; ZFEND; Zfp358; Zinc finger protein 358; ZNF358
Gene Symbols ZNF358
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF358 (aa 104-153).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140467
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.