missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF408 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94588-0.1ml
This item is not returnable.
View return policy
Description
ZNF408 Polyclonal antibody specifically detects ZNF408 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| ZNF408 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| A8-51, C2-H2 type zinc finger protein, FLJ20216, HF.12, HZF3.1, KOX25zinc finger protein 3 (A8-51), PP838, Zfp113, zinc finger protein 3, Zinc finger protein HF.12, Zinc finger protein HZF3.1, Zinc finger protein KOX25 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human ZNF408 (NP_079017.1). MEEAEELLLEGKKALQLAREPRLGLDLGWNPSGEGCTQGLKDVPPEPTRDILALKSLPRGLALGPSLAKEQRLGVWCVGDPLQPGLLWGPLEEESASKEKGEGVKPRQEENLSLGPWGDVCACEQSSGWTSLVQRGRLESEGNVAPVRISERLHLQVYQLVLPGSELLLWPQPSSEGPSLTQPGLDKEAAVAVVTEVESAVQQEVASPGEDAAEPCIDPGSQSPSGIQAENMVSPGLKFP | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79797 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction