missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF415 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15564902
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15564902 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15564902 Supplier Invitrogen™ Supplier No. PA541594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: MAFTQLTFRD VAIEFSQDEW KCLNSTQRTL YRDVMLENYR NLVSLDLSRN Sequence homology: Cow: 93%; Horse: 93%; Human: 100%; Mouse: 100%; Pig: 93%; Rat: 100%; Sheep: 93%.

ZNF415 is involved in transcriptional regulation. The transcriptional activity differed among the various isoforms. All isoforms except isoform 3 seem to suppress the transcriptional activities of AP-1 and p53.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF415
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF415
Gene Accession No. Q09FC8
Gene Alias ZfLp; Zinc finger protein 415; ZNF415
Gene Symbols ZNF415
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human ZNF415.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55786
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.