missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF44 Monoclonal Antibody (4E2)
GREENER_CHOICE

Product Code. 15549062
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15549062 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15549062 Supplier Invitrogen™ Supplier No. MA521976

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Peptide Sequence: MALCYGTFWG YPKMLEAANL MEGLVDIGPW VTLPRGQPEV LEWGLPKDQD SVAFEDVAVN FTHEEWALLG PSQKNLYRDV MRETIRNLNC IGMKWENQNI DDQ.

ZNF44 may be involved in transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF44
Applications ELISA, Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 4E2
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene ZNF44
Gene Accession No. P15621
Gene Alias GIOT2; GIOT-2; gonadotropin inducible transcription repressor-2; gonadotropin-inducible ovary transcription repressor 2; gonadotropin-inducible transcription repressor 2; KOX7; Zinc finger protein 44; zinc finger protein 55; Zinc finger protein 58; zinc finger protein KOX7; zinc finger protein ZnFP12; ZNF; ZNF44; ZNF504; ZNF55; ZNF58
Gene Symbols ZNF44
Host Species Mouse
Immunogen ZNF44 (NP_057348.2, 1 a.a. ~ 103 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51710
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.