missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF500 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
198.00 € - 468.00 €
Specifications
| Antigen | ZNF500 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ZNF500 Polyclonal antibody specifically detects ZNF500 in Mouse samples. It is validated for Western BlotSpecifications
| ZNF500 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| C2H2-171, RP58Zinc finger and BTB domain-containing protein 18, TAZ1, TAZ-1Zinc finger protein C2H2-171, Transcriptional repressor RP58, Translin-associated zinc finger protein 1, ZBTB1858 kDa repressor protein, zinc finger protein 238 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ZNF500 (NP_067678.1). MATVPGLQPLPTLEQDLEQEEILIVKVEEDFCLEEEPSVETEDPSPETFRQLFRLFCYQEVAGPREALSRLWELCCRWLRPELRTKEQILELLVLEQFLTVLPGEIQARVREQQPESGEEAVVLVEGLQRKPRKHRQRGSELLSDDEVPLGIGGQFLKHQAEAQPEDLSLEEEARFSSQQPPAQLSHRPQRGPLLWPERGPPAPRHQEMASASPFLSAWSQVPVNLEDVAVYLSGEEPRCMDPAQRDAPLENEGPGIQLEDGGDGREDAPLRMEWYRVLSARCQGPGHPLPGQRPAPVRG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 26048 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title