missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF500 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94358-0.02ml
This item is not returnable.
View return policy
Description
ZNF500 Polyclonal antibody specifically detects ZNF500 in Mouse samples. It is validated for Western Blot
Specifications
| ZNF500 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| C2H2-171, RP58Zinc finger and BTB domain-containing protein 18, TAZ1, TAZ-1Zinc finger protein C2H2-171, Transcriptional repressor RP58, Translin-associated zinc finger protein 1, ZBTB1858 kDa repressor protein, zinc finger protein 238 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ZNF500 (NP_067678.1). MATVPGLQPLPTLEQDLEQEEILIVKVEEDFCLEEEPSVETEDPSPETFRQLFRLFCYQEVAGPREALSRLWELCCRWLRPELRTKEQILELLVLEQFLTVLPGEIQARVREQQPESGEEAVVLVEGLQRKPRKHRQRGSELLSDDEVPLGIGGQFLKHQAEAQPEDLSLEEEARFSSQQPPAQLSHRPQRGPLLWPERGPPAPRHQEMASASPFLSAWSQVPVNLEDVAVYLSGEEPRCMDPAQRDAPLENEGPGIQLEDGGDGREDAPLRMEWYRVLSARCQGPGHPLPGQRPAPVRG | |
| 0.02 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 26048 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction