missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF529 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
483.00 €
Specifications
| Antigen | ZNF529 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF529 Polyclonal specifically detects ZNF529 in Human samples. It is validated for Western Blot.Specifications
| ZNF529 | |
| Polyclonal | |
| Rabbit | |
| NP_066002 | |
| 57711 | |
| Synthetic peptide directed towards the N terminal of human ZNF529. Peptide sequence QMKIISENVPSYKTHESLTLPRRTHDSEKPYEYKEYEKVFSCDLEFDEYQ. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| KIAA1615, zinc finger protein 529 | |
| ZNF529 | |
| IgG | |
| 54 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title