missing translation for 'onlineSavingsMsg'
Learn More

ZNF680 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

466.00 €

Specifications

Antigen ZNF680
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18277226
View Documents
Novus Biologicals
NBP1-79428
100 μL
466.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZNF680 Polyclonal specifically detects ZNF680 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZNF680
Polyclonal
Rabbit
Human
FLJ90430, zinc finger protein 680
ZNF680
IgG
62 kDa
Western Blot
Unconjugated
RUO
NP_848653
340252
Synthetic peptide directed towards the N terminal of human ZNF680. Peptide sequence: RGYGKCGHENLQLRISCKSVDESKVFKEGYNELNQCLRTTQSKIFQCDKY
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.