missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF703 Antibody (CL0654) [Biotin], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52940B
This item is not returnable.
View return policy
Description
ZNF703 Monoclonal antibody specifically detects ZNF703 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
Specifications
| ZNF703 | |
| Monoclonal | |
| Biotin | |
| FLJ14299, ZEPPO1, Zinc finger elbow-related proline domain protein 1, zinc finger protein 703, ZNF503L, ZPO1 | |
| This antibody was developed using a recombinant protein derived from Q9H7S9, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown | |
| CL0654 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 80139 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction