Learn More
ZNF703 Antibody (CL0654) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52940FR
Product Code. 30298324
  • 737.00 € / 0.10mL
Description
ZNF703 Monoclonal antibody specifically detects ZNF703 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZNF703
Monoclonal
DyLight 680
FLJ14299, ZEPPO1, Zinc finger elbow-related proline domain protein 1, zinc finger protein 703, ZNF503L, ZPO1
This antibody was developed using a recombinant protein derived from Q9H7S9, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Human
Purified
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown
CL0654
50mM Sodium Borate
Mouse
Protein A purified
RUO
80139
Store at 4⪚C in the dark.
IgG1