Learn More
ZNF710 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand:  Novus Biologicals H00374655-B01P
Product Code. 18564080
  • 589.00 € / 50µg
Description
ZNF710 Polyclonal antibody specifically detects ZNF710 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ZNF710
Polyclonal
Western Blot 1:500, Immunocytochemistry/ Immunofluorescence
NP_940928
Mouse
Protein A purified
RUO
374655
Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunocytochemistry/Immunofluorescence
Unconjugated
PBS (pH 7.4)
DKFZp547K1113, FLJ00306, FLJ37393, MGC163413, zinc finger protein 710
ZNF710 (NP_940928, 1 a.a. - 664 a.a.) full-length human protein. MEGFMDSGTQTDAVVVLSLAQAAVLGLVSENELFGATISAEAFYPDLGPELSGAAMGEPEPPGPDVYQLACNGRALEEPAEEEVLEVEAACEKHTRRKTRPPVRLVPKVKFEKVEEEEQEVYEVSVPGDDKDAGPAEAPAEAASGGCDALVQSSAVKMIDLSAFSRKPRTLRHLPRTPRPELNVAPYDPHFPAPARDGFPEPSMALPGPEALPTECGFEPPHLAPLSDPEAPSMESPEPVKPEQGFVWQEASEFEADTAGSTVERHKKAQLDRLDINVQIDDSYLVEAGDRQKRWQCRMCEKSYTSKYNLVTHILGHNGIKPHSCPHCSKLFKQPSHLQTHLLTHQGTRPHKCQVCHKAFTQTSHLKRHMLLHSEVKPYSCHFCGRGFAYPSELKAHEVKHESGRCHVCVECGLDFSTLTQLKRHLASHQGPTLYQCLECDKSFHYRSQLQNHMLKHQNVRPFVCTECGMEFSQIHHLKQHSLTHKGVKEFKCEVCGREFTLQANMKRHMLIHTSVRPYQCHICFKTFVQKQTLKTHMIVHSPVKPFKCKVCGKSFNRMYNLLGHMHLHAGSKPFKCPYCSSKFNLKGNLSRHMKVKHGVMDIGLDSQDPMMELTGTDPSELDGQQEMEDFEENAYSYASVDSSAEASVLTEQAMKEMAYYNVL
0.05 mg
Primary
Human
Purified