missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF766 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35621-100ul
This item is not returnable.
View return policy
Description
ZNF766 Polyclonal antibody specifically detects ZNF766 in Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| ZNF766 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA | |
| zinc finger protein 766 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 60-190 of human ZNF766 (NP_001010851.1).,, Sequence:, MMKQRTEPWTVENEMKVAKNPDRWEGIKDINTGRSCAVRSKAGNKPITNQLGLTFQLPLPELEIFQGEGKIYECNQVQKFISHSSSVSPLQRIYSGVKTHIFNKHRNDFVDFPLLSQEQKAHIRRKPYECN | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Purified |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 90321 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion