missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZNF768 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94393-0.1ml
This item is not returnable.
View return policy
Description
ZNF768 Polyclonal antibody specifically detects ZNF768 in Mouse samples. It is validated for Western Blot
Specifications
| ZNF768 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| zinc finger protein 768 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 140-270 of human ZNF768 (NP_078947.3). PGYESESSRYESQNTELKTQSPEFEAQSSKFQEGAEMLLNPEEKSPLNISVGVHPLDSFTQGFGEQPTGDLPIGPPFEMPTGALLSTPQFEMLQNPLGLTGALRGPGRRGGRARGGQGPRPNICGICGKSF | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 79724 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction