missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF830 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15534692
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15534692 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15534692 Supplier Invitrogen™ Supplier No. PA541197

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Peptide sequence: ENTAEALPEG FFDDPEVDAR VRKVDAPKDQ MDKEWDEFQK AMRQVNTISE Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rat: 93%.

Located on chromosome 17, this gene encodes for a coiled-coil domain containing 16 protein with unknown function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF830
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ZNF830
Gene Accession No. Q96NB3
Gene Alias 2410003C20Rik; AV301118; AW048207; CCDC16; coiled-coil domain containing 16; coiled-coil domain-containing protein 16; hypothetical protein MGC20398; MGC20398; Omcg1; orphan maintenance of genome 1; Ovus mutant candidate gene 1 protein; Zfp830; Zinc finger protein 830; Zinc finger protein 830-like protein; Znf830
Gene Symbols ZNF830
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human CCDC16.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 91603
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.