missing translation for 'onlineSavingsMsg'
Learn More

Zonisamide, MedChemExpress

Product Code. 16411386
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
500 mg
Unit Size:
200mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16411386 200 mg 200mg
16402036 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16411386 Supplier Medchem Express Supplier No. HYB0124
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Zonisamide (AD 810) is an inhibitor of zinc enzyme carbonic anhydrase (CA), with Kis of 35.2 nM and 20.6 nM for human mitochondrial isozyme hCA II and hCA V, respectively. Zonisamide has antiepileptic activity. Zonisamide can be used for the rsearch for epilepsy, seizures and Parkinson's disease.

Zonisamide (AD 810) is an inhibitor of zinc enzyme carbonic anhydrase (CA), with Kis of 35.2 nM and 20.6 nM for human mitochondrial isozyme hCA II and hCA V, respectively. Zonisamide has antiepileptic activity. Zonisamide can be used for the rsearch for epilepsy, seizures and Parkinson's disease.

Specifications

Chemical Name or Material Zonisamide
CAS 68291-97-4
Color White
Purity Grade Notes Research
Molecular Formula C8H8N2O3S
Quantity 200 mg
Synonym AD 810 CI 912
Solubility Information DMSO : 250 mg/mL (1177.97 mM; Need ultrasonic) ∣H2O : 0.67 mg/mL (3.16 mM; Need ultrasonic)
SMILES O=S(CC1=NOC2=C1C=CC=C2)(N)=O
Molecular Weight (g/mol) 212.23
Formula Weight 212.23
Percent Purity 99.85%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.