missing translation for 'onlineSavingsMsg'
Learn More

Zosuquidar trihydrochloride, MedChemExpress

Product Code. 16458078
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458078 10 mg 10mg
16457846 100 mg 100mg
16433997 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16458078 Supplier Medchem Express Supplier No. HY50671
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Zosuquidar (RS 33295-198) trihydrochloride is an inhibitor of P-glycoprotein with a Ki value of 59 nM.

Zosuquidar (RS 33295-198) trihydrochloride is an inhibitor of P-glycoprotein with a Ki value of 59 nM.

Specifications

Chemical Name or Material Zosuquidar trihydrochloride
CAS 167465-36-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C32H34Cl3F2N3O2
Quantity 10 mg
Synonym RS 33295-198 trihydrochloride LY-335979 trihydrochloride
Solubility Information H2O : 5 mg/mL (7.85 mM; Need ultrasonic) ∣DMSO : 1 mg/mL (1.57 mM; Need ultrasonic)
SMILES FC1([C@H]2[C@@H]1C3=C([C@@H](C4=CC=CC=C42)N5CCN(CC5)C[C@H](COC6=C7C=CC=NC7=CC=C6)O)C=CC=C3)F.Cl.Cl.Cl
Molecular Weight (g/mol) 636.99
Formula Weight 636.99
Percent Purity 98.73%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.