missing translation for 'onlineSavingsMsg'
Learn More

ZT 52656A hydrochloride, MedChemExpress

Product Code. 16460789
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
20 mg
Unit Size:
10mg
1mg
20mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16460789 1 mg 1mg
16453438 10 mg 10mg
16430426 20 mg 20mg
16406167 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16460789 Supplier Medchem Express Supplier No. HY101582
 Restricted Item
Estimated Shipment: 11-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress ZT 52656A is a selective kappa opioid agonist, used for the prevention or alleviation of pain in the eye.

ZT 52656A is a selective kappa opioid agonist, used for the prevention or alleviation of pain in the eye.

Specifications

Chemical Name or Material ZT 52656A hydrochloride
Physical Form Solid
CAS 115730-24-0
Color Off-White
Quantity 1 mg
Purity Grade Notes Research
Molecular Formula C19H26ClF3N2O
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 35.71 mg/mL (91.36 mM; Need ultrasonic)
SMILES O=C(CC1=CC=C(C=C1)C(F)(F)F)N2CCCCC2CN3CCCC3.Cl
Molecular Weight (g/mol) 390.87
Formula Weight 390.87
Percent Purity 99.76%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.